Learn More
Medchemexpress LLC Zen-3694 | 1643947-30-1 | 99.7% | 333.39 | C19H19N5O | 25MG
SDP
Catalog No. 5000235940
Encompass_Preferred
BET-IN-19 is a potent small-molecule BET bromodomain inhibitor for research use. It inhibits hIL-6 mRNA transcription and c-myc activity in AML MV4-11 cells (IC50 ≤ 0.3 μM). The compound has molecular formula C19H19N5O, molecular weight 333.39, and reported purity ≈99.7%; it is supplied as a powder and is typically stored at -20°C (powder) or at -80°C in solvent for longer term.
Catalog No. 50-002-35940 Supplier Medchemexpress LLC Supplier No. HY12523625MG
BET-IN-19 is a potent small-molecule BET bromodomain inhibitor for research use. It inhibits hIL-6 mRNA transcription and c-myc activity in AML MV4-11 cells (IC50 ≤ 0.3 μM). The compound has molecular formula C19H19N5O, molecular weight 333.39, and reported purity ≈99.7%; it is supplied as a powder and is typically stored at -20°C (powder) or at -80°C in solvent for longer term.