Learn More
Selleck Chemical LLC ZEN-3694-E1517-100MG
SDP
Catalog No. 503134808
Encompass_Preferred
ZEN-3694 is an orally bioavailable bromodomain extraterminal inhibitor (BETi) that leads to down-regulation of expression of AR-signaling in metastatic castrationresistant prostate cancer (mCRPC) models
Catalog No. 50-313-4808 Supplier Selleck Chemical LLC Supplier No. E1517100MG
ZEN-3694 is an orally bioavailable bromodomain extraterminal inhibitor (BETi) that leads to down-regulation of expression of AR-signaling in metastatic castrationresistant prostate cancer (mCRPC) models