Learn More
Selleck Chemical LLC ZEN-3694-E1517-5MG
SDP
Supplier:  Selleck Chemical LLC E15175MG
Encompass_Preferred
ZEN-3694 is an orally bioavailable bromodomain extraterminal inhibitor (BETi) that leads to down-regulation of expression of AR-signaling in metastatic castrationresistant prostate cancer (mCRPC) models
Catalog No. 50-313-7236