Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp ZenFit Assembly, Capillary, 40μm, 6 in., HP

Catalog No. 502391646 Shop All Waters Corporation Products
Encompass_Preferred

Replacement ZenFit Assembly, Capillary and Fittings, 40μm x 6 in., HP.

  • Product type: Spare parts kits
  • Type: Mechanical
  • System type: UPLC
  • Replacement schedule: As required

Catalog No. 50-239-1646 Supplier Waters Corp Supplier No. 700011468
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement ZenFit Assembly, Capillary and Fittings, 40μm x 6 in., HP.

  • Product type: Spare parts kits
  • Type: Mechanical
  • System type: UPLC
  • Replacement schedule: As required

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.