Learn More
Waters Corp Waters Corporation ZenFit Assembly, Capillary and Fittings, 40μm, 26 in.
Catalog No. 502039837 Shop All Waters Corporation Products
Encompass_Preferred
The Waters ZenFit Assembly, Capillary and Fittings provides a replacement solution for UPLC systems. This mechanical assembly includes a 40μm capillary that is 26 in. long, ensuring optimal performance for your instrumentation.
  • Replacement part for UPLC systems.
  • Mechanical type.
  • Includes capillary and fittings.
  • Features a 40μm capillary.
  • 26 in. length.
  • Replace as required.
Catalog No. 50-203-9837 Supplier Waters Corp Supplier No. 700011469
The Waters ZenFit Assembly, Capillary and Fittings provides a replacement solution for UPLC systems. This mechanical assembly includes a 40μm capillary that is 26 in. long, ensuring optimal performance for your instrumentation.
  • Replacement part for UPLC systems.
  • Mechanical type.
  • Includes capillary and fittings.
  • Features a 40μm capillary.
  • 26 in. length.
  • Replace as required.