Learn More
Waters Corp ZenFit Assembly, Finger-Tight, Re-usable, ionKey Source
Catalog No. 502391000 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ZenFit Connection Technology provides re-usable, finger-tight connectors for ACQUITY UPLC M-Class Systems, enabling tool-free installation. This technology prevents poor connections, maintains flow path integrity, and reduces chromatographic variability. It withstands pressures up to 15,000 psi and temperatures up to 90 °C, making nano- and micro-scale UPLC more accessible.
  • Re-usable finger-tight fittings.
  • 15,000 psi pressure tolerance.
  • Up to 90 °C temperature tolerance.
  • Enhances mass spectrometry sensitivity by increasing sampling efficiency, improving ionization, and reducing matrix effects.
  • Compatible with ACQUITY UPLC M-Class system.
  • Compatible with HDX technology.
  • Compatible with ionKey/MS system.
  • Compatible with nanoEase M/Z columns.
Catalog No. 50-239-1000 Supplier Waters Corp Supplier No. 700011463
ZenFit Connection Technology provides re-usable, finger-tight connectors for ACQUITY UPLC M-Class Systems, enabling tool-free installation. This technology prevents poor connections, maintains flow path integrity, and reduces chromatographic variability. It withstands pressures up to 15,000 psi and temperatures up to 90 °C, making nano- and micro-scale UPLC more accessible.
  • Re-usable finger-tight fittings.
  • 15,000 psi pressure tolerance.
  • Up to 90 °C temperature tolerance.
  • Enhances mass spectrometry sensitivity by increasing sampling efficiency, improving ionization, and reducing matrix effects.
  • Compatible with ACQUITY UPLC M-Class system.
  • Compatible with HDX technology.
  • Compatible with ionKey/MS system.
  • Compatible with nanoEase M/Z columns.