Learn More
Medchemexpress LLC Zenidolol hydrochloride | 72795-01-8 | 99.7% | 313.86 | 1 ML
SDP
Catalog No. 5000350184
Encompass_Preferred
Zenidolol hydrochloride (ICI-118551 hydrochloride) is a highly selective β2 adrenergic receptor antagonist. It exhibits Kis of 0.7, 49.5, and 611 nM for β2, β1, and β3 receptors, respectively, making it a potent research tool for studying adrenergic receptor activity.
  • Inhibits cAMP accumulation with an IC50 of 1.7 μM in IMCD cells.
  • Induces prominent vasorelaxation of norepinephrine pre-contracted PA.
  • Significantly affects beat duration in failing human heart.
  • Reduces systolic pressure in the pulmonary circuit in mice.
  • Negative inotropic effect not cAMP-related.
  • Available in solid, solution, or solid + solvent formats.
Catalog No. 50-003-50184 Supplier Medchemexpress LLC Supplier No. HY1395110MM/1ML
Zenidolol hydrochloride (ICI-118551 hydrochloride) is a highly selective β2 adrenergic receptor antagonist. It exhibits Kis of 0.7, 49.5, and 611 nM for β2, β1, and β3 receptors, respectively, making it a potent research tool for studying adrenergic receptor activity.
  • Inhibits cAMP accumulation with an IC50 of 1.7 μM in IMCD cells.
  • Induces prominent vasorelaxation of norepinephrine pre-contracted PA.
  • Significantly affects beat duration in failing human heart.
  • Reduces systolic pressure in the pulmonary circuit in mice.
  • Negative inotropic effect not cAMP-related.
  • Available in solid, solution, or solid + solvent formats.