Learn More
Medchemexpress LLC Zenidolol hydrochloride | 72795-01-8 | 99.7% | 313.86 | 100 MG
SDP
Catalog No. 5000364951
Encompass_Preferred
Zenidolol hydrochloride (ICI-118551) is a highly selective β2 adrenergic receptor antagonist, demonstrating potent inhibition with Kis of 0.7, 49.5, and 611 nM for β2, β1, and β3 receptors, respectively. It is for research use only.
  • Highly selective β2 adrenergic receptor antagonist
  • Inhibits cAMP accumulation with an IC50 of 1.7 μM in IMCD cells
  • Induces prominent vasorelaxation of norepinephrine (NE)-precontracted PA but not AO (at 10 μM)
  • Has significant effects on beat duration in failing human heart, reducing time-to-peak contraction and time-to-90% relaxation compared with basal contraction
  • Negative inotropic effect is not cAMP-related
  • Overexpression of β2AR in rabbit myocytes enhances negative inotropic effects
  • Reduces systolic pressure in the pulmonary circuit but not systemic arterial pressure when injected into the jugular vein of mice (at 0.2 mg/kg)
Catalog No. 50-003-64951 Supplier Medchemexpress LLC Supplier No. 100MGHY13951
Zenidolol hydrochloride (ICI-118551) is a highly selective β2 adrenergic receptor antagonist, demonstrating potent inhibition with Kis of 0.7, 49.5, and 611 nM for β2, β1, and β3 receptors, respectively. It is for research use only.
  • Highly selective β2 adrenergic receptor antagonist
  • Inhibits cAMP accumulation with an IC50 of 1.7 μM in IMCD cells
  • Induces prominent vasorelaxation of norepinephrine (NE)-precontracted PA but not AO (at 10 μM)
  • Has significant effects on beat duration in failing human heart, reducing time-to-peak contraction and time-to-90% relaxation compared with basal contraction
  • Negative inotropic effect is not cAMP-related
  • Overexpression of β2AR in rabbit myocytes enhances negative inotropic effects
  • Reduces systolic pressure in the pulmonary circuit but not systemic arterial pressure when injected into the jugular vein of mice (at 0.2 mg/kg)