Learn More
Med Vet International Zoetis Zeniquin Tablets, 100mg, 50ct.
Catalog No. 502828938
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Zeniquin (marbofloxacin) tablets are indicated for the treatment of infections associated with bacteria susceptible to marbofloxacin, including bacterial skin and soft tissue infections in dogs and cats and urinary tract infections (cystitis) in dogs. For oral use in dogs and cats only. Federal law prohibits the extra-label use of this drug in food-producing animals.
Catalog No. 50-282-8938 Supplier Med Vet International Supplier No. RXZEN10050
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Zeniquin (marbofloxacin) tablets are indicated for the treatment of infections associated with bacteria susceptible to marbofloxacin, including bacterial skin and soft tissue infections in dogs and cats and urinary tract infections (cystitis) in dogs. For oral use in dogs and cats only. Federal law prohibits the extra-label use of this drug in food-producing animals.