Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zenon™ Human IgG Labeling Kits

Catalog No. Z25408
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 Reactions kit
50 Reactions kit
Excitation/Emission:
495/519 nm
496, 546, 565/578 nm
590/617 nm
650/660 nm
650/668 nm
Label or Dye:
APC (Allophycocyanin)
Alexa Fluor 488
Alexa Fluor 594
R-PE (R-Phycoerythrin)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Excitation/Emission Label or Dye
Z25408 50 Reactions kit 650/668 nm -
Z25402 50 Reactions kit 495/519 nm Alexa Fluor 488
Z25407 50 Reactions kit 590/617 nm Alexa Fluor 594
50-113-7626 25 Reactions kit 650/660 nm APC (Allophycocyanin)
50-591-315 25 Reactions kit 496, 546, 565/578 nm R-PE (R-Phycoerythrin)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. Z25408 Supplier Invitrogen™ Supplier No. Z25408
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Contains 1 vial of Zenon Alexa Fluor 647 Human IgG labeling reagent (250 μL), and 1 vial of Zenon blocking reagent (rabbit IgG, 250 μL).

Take advantage of a fast, versatile, and reliable method for producing antibody (IgG) conjugates for immunocytochemistry (ICC), immunohistochemistry (IHC), flow cytometry, and cell imaging with the Invitrogen Zenon™ Human IgG Labeling Kits.

Take advantage of a fast, versatile, and reliable method for producing antibody (IgG) conjugates for immunocytochemistry (ICC), immunohistochemistry (IHC), flow cytometry, and cell imaging with the Invitrogen Zenon™ Human IgG Labeling Kits. Simplify your laboratory applications and reduce antibody cross-reactivity while also achieving efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions.

Achieve fast, versatile, and reliable fluorophore-, biotin-, or enzyme-labeled IgG primary antibodies with the Invitrogen Zenon™ Human IgG Labeling Kits. These kits utilize Alexa Fluor fluorophores, biotin, or enzymes such as R-phycoerythrin and allophycocyanin, which are attached to monovalent, affinity purified Fab fragments. The Fab fragments, in turn, are directed against and bind with the Fc portion of IgG primary antibodies. Only a small amount of starting material is required, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions. Because the Zenon labeling method is based on immunoselectivity, it does not require the removal of exogenous proteins (such as serum) or amine-containing buffers from the target antibody, simplifying the process.

Important features of Zenon labeling technology:
• Labeled antibodies are typically ready to use in 10 minutes
• Requires only 1–20 μg primary antibody
• Simple—no purification required
• Flexible—choose from different fluorophores, biotin, or enzymes.
• Versatile—can be used in a variety of applications including ICC, IHC, flow cytometry, and cell imaging.

Advantages of using Zenon antibody labeling kits:

Cost savings
Zenon antibody labeling kits offer a cost-conscious and reproducible method of tagging as little as 0.4 μg in 2 μL of primary antibody, with minimal waste of expensive or difficult-to-obtain antibodies, or excessive washing steps that pose the risk of product loss.

Sensitivity
Label your primary antibodies without compromising their antigen binding affinity: Zenon dye- and enzyme-labeled Fab fragments, which are targeted to the Fc tail, are affinity purified during their preparation to ensure high affinity and selectivity for the Fc portion of the corresponding primary antibody. The procedure for chemical labeling of the Zenon Fab fragments protects their Fc-binding site, resulting in more active labeling reagents.

Speed
No purification procedure is required prior to using Zenon Fab fragments in your laboratory applications. Formation of the Fab-antibody complex occurs in fewer than five minutes, followed by a five-minute blocking step. During this time, almost all the primary antibody in the mixture is labeled with the labeled Fab fragments.

Simplicity
The Fab-antibody complexes display fluorescence or enzymatic activity that is similar in intensity to that of directly labeled primary antibodies. Varying the extent of the antibody labeling is as simple as changing the amount of added Zenon labeling reagent during the reaction. Once the labeling complexes are formed, they can used immediately, without need for antibody purification.

Reliability
The Zenon Fab-antibody complex is stable and allows subsequent or simultaneous labeling of different target cells and tissues with different complexes. After staining, an aldehyde-based fixing step may be used to prevent the transfer of different labels between different primary antibodies, preserving the initial staining pattern.

Customization
We offer custom antibody conjugation services that are efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Labeling Kit
Quantity 50 Reactions kit
Content And Storage Contains 1 vial of Zenon Alexa Fluor 647 Human IgG labeling reagent (250 μL), and 1 vial of Zenon blocking reagent (human IgG, 250 μL).

Store in refrigerator (2–8°C) and protect from light.
Excitation/Emission 650/668 nm
Labeling Scale < 1–20 μg
Product Line Zenon
Color Far-Red
Labeling Target IgG
Species Human
Label Type Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.