Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zenon™ LysoLight™ Green IgG Labeling Reagents

Catalog No. Z25628
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μL
1.2 mL
Label Type:
Human IgG Fc Domain
Mouse IgG Fc Domain
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Label Type
Z25628 1.2 mL Human IgG Fc Domain
Z25627 250 μL Human IgG Fc Domain
Z25629 250 μL Mouse IgG Fc Domain
Z25630 1.2 mL Mouse IgG Fc Domain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. Z25628 Supplier Invitrogen™ Supplier No. Z25628
Only null left
Add to Cart
Add to Cart

Streamline antibody-drug conjugate screening with Zenon LysoLight Green IgG labeling reagents for the monitoring of antibody internalization. These reagents offer rapid, scalable antibody labeling and a bright fluorescence-based indication of antibody uptake and lysosomal trafficking in target cells when analyzed by flow cytometry, fluorescence microscopy, or high-content screening.

Streamline antibody-drug conjugate screening with the use of Invitrogen Zenon LysoLight Green IgG labeling reagents for the monitoring of antibody internalization. These reagents offer rapid, scalable antibody labeling and a bright fluorescence-based indication of antibody uptake and lysosomal trafficking in target cells when analyzed by flow cytometry, fluorescence microscopy, or high-content screening.

Features of Zenon LysoLight Green IgG labeling reagents include:

  • Simplicity—no reagent purification is required
  • Accuracy—no wash steps or quenchers are needed, minimizing cross-reactivity or background
  • Speed—labeled antibodies are typically ready to use in 10 minutes
  • Specificity—dyes are designed for the specific detection of lysosomal trafficking

Zenon LysoLight Green-labeled Fab fragments (i.e., labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex. Formation of the Fab–antibody complex occurs in fewer than five minutes. Because the LysoLight dye fluorescence dramatically increases upon lysosomal catabolism, the antibodies coupled with the Zenon LysoLight Green IgG labeling reagent provide an excellent measure of lysosomal delivery of internalized antibodies.

Zenon LysoLight Green IgG labeling reagents contain sufficient material to label up to sixteen 96-well plates, depending on the product size and Zenon labeling reagent concentration used. Only a small amount of starting material is needed, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions.

Preserve primary antibody function and affinities

Reactive dye labeling of primary antibodies can have unpredictable and undesirable outcomes. One of these is reduced binding affinity due to label addition in the binding pocket. The Zenon antibody labeling approach, which is targeted to the Fc tail, resolves this potential issue.

Try custom antibody conjugates

If the antibody conjugate you need is not available as one of our stocked products, we will prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) To screen for antibody internalization
Product Type Labeling Reagent
Quantity 1.2 mL
Sufficient For 4 to 16 96-well plates
Content And Storage 2°C to 8°C
Detection Method Fluorescence
Excitation/Emission 488/525 nm
For Use With (Equipment) Flow Cytometer, Fluorescence Microscope, High Content Analysis Instrument
Product Line LysoLight
Chemical Reactivity High Affinity
Solubility Aqueous Buffer
Label or Dye LysoLight Green
Spacer None
Color Green
Labeling Target Antibodies
Flow Cytometer Laser Lines Blue
Label Type Human IgG Fc Domain
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.