Learn More
Invivogen Zeocin Selective antibiotic for the Sh ble gene, 1g (10 x 1 ml)
SDP
Catalog No. NC9002627
Encompass
Zeocin, 1 g Liquid @ 100 mg/mL, Color: Blue, Storage: -20 deg.C, Purity: 90% > HPLC and Microbiological Assays, Packaging: 5 x 2 mL Tubes, pH: 7.25, CAS No: 11006-33-0
Catalog No. NC9002627 Supplier Invivogen Supplier No. ANTZN1
Zeocin, 1 g Liquid @ 100 mg/mL, Color: Blue, Storage: -20 deg.C, Purity: 90% > HPLC and Microbiological Assays, Packaging: 5 x 2 mL Tubes, pH: 7.25, CAS No: 11006-33-0