Learn More
Invivogen ZEOCIN 5G 50 ML BOTTLE
SDP
Catalog No. NC9497748
Encompass
Zeocin (Phleomycin); 50mL bottle; Unconjugated; Is a formulation of Phleomycin D1, Copper-Chelated Glycopeptide Antibiotic produced by Streptomyces CL990
Catalog No. NC9497748 Supplier Invivogen Supplier No. ANTZN5B
Zeocin (Phleomycin); 50mL bottle; Unconjugated; Is a formulation of Phleomycin D1, Copper-Chelated Glycopeptide Antibiotic produced by Streptomyces CL990