Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Zeocin™ Selection Reagent

Catalog No. r25001
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
8 x 1.25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R25001 8 x 1.25 mL
R25005 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R25001 Supplier Gibco™ Supplier No. R25001
Only null left
Add to Cart
Edge
Add to Cart

Zeocin Selection Antibiotic is a formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus.

Gibco Zeocin Selection Antibiotic is a formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus. It acts by binding to and cleaving DNA, leading to cell death, and is used for selection of of mammalian, insect, yeast, bacterial, and plant cell cultures. Working concentration range is 50–400 μg/mL depending on the cell type. This product is supplied at 100 mg/mL in sterile water in 8 x 1.25 mL vials.

  • Gibco™ Zeocin™ Selection Antibiotic is a formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus
  • Zeocin™ Selection Antibiotic acts by binding to and cleaving DNA, leading to cell death
  • It is used for selection of of mammalian, insect, yeast, bacterial, and plant cell cultures
  • Working concentration ranges between 50–400 μg/mL depending on the cell type
  • This product is supplied at 100 mg/mL in sterile water, in 8 x 1.25 mL vials

Order Info

Shipping conditions: Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -10 to -30°C
Shipping conditions: Ice
Concentration 100 mg/mL
Culture Type Mammalian Cell Culture, Insect Cell Culture
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
For Use With (Application) Eukaryotic Selection/Stable Cell Line Generation, Bacterial Selection
Product Line Zeocin
Quantity 8 x 1.25 mL
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.