Learn More
Enterprise Technology Solutions Zep Commercial Floor Cleaner/Stripper,Liquid,Biodegradable,Citrus,OE
SDP
Catalog No. 5000810577
Encompass_Preferred
EZ Floor Cleaner/Wax Stripper provides floor maintenance and general-purpose cleaning. Controlled foam easily rinses and avoids suds buildup. Biodegradable formula dilutes effortlessly in water and leaves behind a film-free surface. Cleaner performs equally as well as a floor/hard-surface cleaner or as a wax/finish stripper. Formula contains no ammonia and is inhibited to protect soft metals such as aluminum. Leave behind a clean or finished surface with a pleasant, citrus scent.
Catalog No. 50-008-10577 Supplier Enterprise Technology Solutions Supplier No. ZPE77885
EZ Floor Cleaner/Wax Stripper provides floor maintenance and general-purpose cleaning. Controlled foam easily rinses and avoids suds buildup. Biodegradable formula dilutes effortlessly in water and leaves behind a film-free surface. Cleaner performs equally as well as a floor/hard-surface cleaner or as a wax/finish stripper. Formula contains no ammonia and is inhibited to protect soft metals such as aluminum. Leave behind a clean or finished surface with a pleasant, citrus scent.