Learn More
MSC ZEP Commercial Liquid Bathroom Cleaner, Chlorine Scent, 1 Gallon, Disinfectant
Catalog No. 502838920
Encompass
ZEP Commercial liquid bathroom cleaner is designed for effective cleaning and disinfecting. This 1 gallon bottle features a chlorine scent and is suitable for use on tile, tubs, and showers, making it ideal for various bathroom cleaning tasks.
  • Chlorine scent for a fresh clean smell
  • Functions as a powerful disinfectant
  • Ideal for cleaning tile, tubs, and showers
  • Convenient bottle container type
  • Liquid form for easy application
  • Contains 1 gallon of cleaner
  • Primary chemical is sodium hypochlorite
  • Suitable for use with toilet bowls, sinks, unpainted surfaces, ceramic tile, and grout
Catalog No. 50-283-8920 Supplier MSC Supplier No. 54491352
ZEP Commercial liquid bathroom cleaner is designed for effective cleaning and disinfecting. This 1 gallon bottle features a chlorine scent and is suitable for use on tile, tubs, and showers, making it ideal for various bathroom cleaning tasks.
  • Chlorine scent for a fresh clean smell
  • Functions as a powerful disinfectant
  • Ideal for cleaning tile, tubs, and showers
  • Convenient bottle container type
  • Liquid form for easy application
  • Contains 1 gallon of cleaner
  • Primary chemical is sodium hypochlorite
  • Suitable for use with toilet bowls, sinks, unpainted surfaces, ceramic tile, and grout