Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC ZEP Car Wash, Concentrated, Polymer Wax, Pleasant Scent, 55 gal

Catalog No. 502816925
Encompass

Zep-O-Shine Concentrated Car Wash is a liquid vehicle wash designed for transportation cleaning. This concentrated formula includes polymer wax and delivers a pleasant scent, suitable for professional vehicle maintenance.

  • Formulated as a liquid vehicle wash
  • Supplied in a 55 gal drum
  • Features a pleasant scent and pink color
  • Includes polymer wax for enhanced cleaning
  • Has a pH range of 6.39–7.0
  • Density of 1.028 g/cm³
  • Soluble in water
  • Kinematic viscosity of 125.7 mm²/s at 20 °C

Catalog No. 50-281-6925 Supplier MSC Supplier No. 17732645
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zep-O-Shine Concentrated Car Wash is a liquid vehicle wash designed for transportation cleaning. This concentrated formula includes polymer wax and delivers a pleasant scent, suitable for professional vehicle maintenance.

  • Formulated as a liquid vehicle wash
  • Supplied in a 55 gal drum
  • Features a pleasant scent and pink color
  • Includes polymer wax for enhanced cleaning
  • Has a pH range of 6.39–7.0
  • Density of 1.028 g/cm³
  • Soluble in water
  • Kinematic viscosity of 125.7 mm²/s at 20 °C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.