Learn More
Biolog Inc Anaerobic Chamber Cleaner - 1 Gal
SDP
Catalog No. NC0195813
Encompass
Chamber Cleaner is a surface cleaner for use inside anaerobic chambers or glove boxes. Chamber Cleaner is a 0.13% solution of benzalkonium chloride. Benzalkonium chloride is a quaternary ammonium compound and cationic surfactant. Quaternary ammonium compounds mode of action is irreversibly binding to the phospholipids and proteins of the membrane, thereby impairing permeability.
This item is not returnable. View return policy
Catalog No. NC0195813 Supplier Biolog Inc Supplier No. AS972
Chamber Cleaner is a surface cleaner for use inside anaerobic chambers or glove boxes. Chamber Cleaner is a 0.13% solution of benzalkonium chloride. Benzalkonium chloride is a quaternary ammonium compound and cationic surfactant. Quaternary ammonium compounds mode of action is irreversibly binding to the phospholipids and proteins of the membrane, thereby impairing permeability.