Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Candida albicans Z006, DNA
SDP

Catalog No. 21020101 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-020-101 1 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-020-101 Supplier ZeptoMetrix Supplier No. 0801504DNA1UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for use as an amplification and/or detection control for nucleic acid testing of Candida albicans

  • Identification of this organism was confirmed by rDNA sequencing
  • Purity of the culture was monitored by additional culturing and Gram staining
  • Extracted DNA also tested positive on an in-house real-time PCR assay

Specifications

Content And Storage Store at -20°C.
Formulation Supplied in TE Buffer
For Use With (Application) Amplification and/or detection control for nucleic acid testing of Candida albicans; determination of a limit of detection (LOD), in diagnostic assay development, cross-reactivitystudies or genomic sequencing
Quantity 1 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.