Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Chicken IgY ELISA
SDP

Catalog No. 21020100 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
96 Determinations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-020-100 96 Determinations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-020-100 Supplier ZeptoMetrix Supplier No. 0801010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A rapid, easy-to-use ELISA designed for the measurement of chicken IgY in chicken serum, egg yolk or white. Plasma, saliva, mucosa, cell culture fluid from chicken cells or other biological samples

  • Chicken IgY (IgG) ELISA is designed to measure chicken IgY in serum, egg yolk/white, plasma, saliva, mucosa, and cell culture fluids. Key specifications and features include:
    • Conisists of 96-well microplate (coated with goat anti-chicken IgY)
    • Detection Method: HRP-conjugated goat anti-chicken IgY detector antibody (Fc-fragment specific)
    • Standard: Provided at 125 ng/mL
    • Measurement: Spectrophotometric reading at 450 nm
    • Specificity: Reacts with Fc portion of chicken IgY and no cross-reactivity with non-immunoglobulin proteins or serum from sheep, rabbit, rat, donkey, goat, horse, bovine, mouse, or human
    • Precision: Intra-assay CV 3-7%; Inter-assay CV 5-10%
    • Regulatory Status: Research use only (RUO)
    • Storage: 2°C to 8°C (do not freeze)

Specifications

Chromagen Type Tetramethyl Benzidine (TMB)
Conjugate HRP
Content And Storage Store all kit reagents at 2° to 8°C. Do not freeze. Unused microplate strips should be kept in a sealed bag with desiccant to minimize exposure to moisture.
Product Type Chicken IgY (IgG) ELISA
Quantity 96 Determinations
Species Chicken
For Use With (Application) Measurement of chicken IgY in chicken serum, egg yolk or white, plasma, saliva, mucosa, cell culture fluid from chicken cells or other biological samples
Regulatory Status RUO
Substrate Type TMB
Manufacturing Quality ISO 13485
Includes 96-well microplate, 12 mL detector antibody, 7 mL chicken IgY standard, 100 mL assay diluent, 125 mL 10X plate wash buffer, 12 mL substrate, 12 mL stop solution, 10 microplate sealers, sealable plastic bag, Certificate of Analysis (CoA)
Stability Stable Until the Expiration Date Printed on the Kit Box Label
Target Chicken IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.