Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Coxsackievirus Type A21 Infectious Culture Fluid
SDP

Catalog No. 22157818 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-818 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order
Catalog No. 22-157-818 Supplier ZeptoMetrix Supplier No. 0810235CF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order

For research, product development testing, or quality assurance testing

  • Propagated in the A549 cell line
  • Each lot is assayed for its Tissue Culture Infective Dose (TCID50) and sold with titers >105U/mL

Specifications

Content And Storage -65°C or below
Quantity 1 mL

For research use only. Not for use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.