Learn More
Description
Includes
60mL bottle of ZeptoBind, 120mL bottle of ZeptoBlock, 60mL bottle of ZeptoCoat, 5 microtiter plates per system, 5 sealable bags per system, 5 desiccant pillows per system, 5 plate sealers per system (10 sealers per pack).
- Contains specially screened microtiter plates and all reagents necessary to coat plates with antigen or antibody
- Ensures low background, high sensitivity
- May be stored desiccated for one to two years with no loss of activity
Specifications
Specifications
| Product Type | ELISA construction system |
| Sufficient For | 5 x 96 Microplate Wells |
| For Use With (Application) | The Immuno-Tek ELISA construction System is an easy to use kit that enables scientists to rapidly prepare high quality ELISA assays |
| Kit Contents | 10 sealers/pk., 120 mL Zepto block, 5 per system desiccant pillows, 5 per system microtiter plates, 5 per system plate sealers, 5 per system sealable bags, 60 mL Zepto bind, 60 mL Zepto coat |
| Quantity | 5 x 96 Determinations |
For research purposes or for further manufacturing use only. Not for in vitro diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
