Learn More
Description
Includes
Microplate, (1x96 well), Detector Antibody (12mL), Mouse IgM Standard (7mL), Assay Diluent (100mL), Plate Wash Buffer (125mL), Substrate(12mL), Stop Solution (12mL), Microtiter Plate Sealers (1pk.), Plastic Bag (1 bag).
Requires
Disposable gloves, Test tubes and racks for preparing specimen and IgM standard dilutions, Validated adjustable micropipettes, single and multi-channel, Distilled or deionized water, Validated incubator capable of maintaining 37° ±1°C, Graduated cylinders and assorted beakers, Validated microtiter plate reader, Automatic microtiter plate washer or manual vacuum aspiration equipment, Timer.
- Contains ready-to-use reagents
- Less performance time
Specifications
Specifications
| Product Type | Immuno-Tek Mouse IgM ELISA kit |
| Sufficient For | 96-well Microplate |
| For Use With (Application) | Designed for the measurement of Mouse IgM in Plasma, Serum, hybridoma cell supernatants, ascites or other Mouse biological fluids, the kit is especially useful in monito ring the production and purification of Mouse Monoclonal Antibodies |
| Kit Contents | 1 plastic bag, 100 mL assay diluent, 12 mL detector antibody, 12 mL stop solution, 12 mL substrate, 125 mL plate wash buffer, 7 mL Human IgG standard, 96-well microplate, Microtiter plate sealers |
| Quantity | 96 Determinations |
| Storage Requirements | 2°C to 8°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
