Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ CT/NG External Run Controls
GSA_VASDP

Catalog No. p-4597629 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Format:
Neisseria gonorrhoea detected
No Chlamydia trachomatis and Neisseria gonorrhoea detected
Vial
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
22-157-774 No Chlamydia trachomatis and Neisseria gonorrhoea detected
21-100-544 Vial
21-100-554 Neisseria gonorrhoea detected
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-157-774 Supplier ZeptoMetrix Supplier No. NATCT/NGNEG6MC
Add to Cart
Add to Cart

For determination of the presence of Chlamydia trachomatis and Neisseria gonorrhoeae DNA

  • External run controls formulated with purified, intact and chemically modified organisms
  • Non infectious and refrigerator stable
  • Designed as full process extraction and amplification controls to validate molecular testing
  • Supplied in purified protein matrix that mimics composition of true clinical specimen
  • Evaluates performance of nucleic acid test
  • 6 x 1ml vials (each control)

For verification of clinical assays, development of diagnostic tests and training of laboratory personnel

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Format No Chlamydia trachomatis and Neisseria gonorrhoea detected
Media Type Vial
Product Type External Run Control
Formulation Formulated with purified, intact bacterial cells
For Use With (Application) Quality control of clinical assays and training of laboratory personnel
Quantity 6 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.