Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ HIV-1 External Run Controls, IIIB Strain
SDP

Catalog No. p-4597641 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Concentration:
1000IU/mL
50000IU/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
22-156-894 1000IU/mL
22-156-895 50000IU/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-156-894 Supplier ZeptoMetrix Supplier No. NATHIV1ERCL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to evaluate the performance of nucleic acid tests for determination of the presence of HIV-1 RNA

  • Formulated with purified, intact virus particles that are chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
  • Contains 6 x 1mL vials
  • Available concentrations: 1000 or 50,000 IU/mL

NATHIV1-ERCL and NATHIV1-ERCM contain intact virus and should be run in a manner identical to that used for clinical specimens

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Certifications/Compliance ISO, 13485
Concentration 1000IU/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type External Run Control
Formulation Formulated with purified, intact virus particles
For Use With (Application) Determination of presence of HIV-1 RNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 6 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.