Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Clostridium difficile Verification Panel
GSA_VASDP

Catalog No. 22156714 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-714 6 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-714 Supplier ZeptoMetrix Supplier No. NATCDIVPC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulated with purified, intact chemically modified bacterial particles to render them non-infectious and refrigerator stable

  • contains 6 x 1.0mL vials each containing C. difficile or C.sordellii NATtrol™
  • Supplied in purified protein matrix identical to composition of true clinical specimen

Panel members:

  • C.difficile, Strain: Ribotype 002
  • C.difficile, Strain: Ribotype 017
  • C.difficile, Strain: Ribotype 027
  • C.difficile, Strain: Ribotype 027
  • C.sordellii
  • Verification of clinical assays, development of diagnostic tests and training of laboratory personnel

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance ISO 9001:2008 and ISO 13485:2003 Standards
Content And Storage 2°C to 8°C
Type Bacteria Verification Panel
Includes C. difficile ribotype 002, 078, 017, 027, C. sordellii
Intended Use Verification of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 6 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.