Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Mycoplasma pneumoniae External Run Control
SDP

Catalog No. 22157799 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Concentration:
50,000 copies/mL
5000 copies/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
22-157-799 50,000 copies/mL
22-157-798 5000 copies/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-157-799 Supplier ZeptoMetrix Supplier No. NATMPN(M129)ERCM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulated with intact chemically modified bacteria to render them non-infectious and refrigerator stable

  • Each run control pack contains 6 x 1mL vials of
      M. pneumoniae
    NATtrol™ at concentrations of 5,000 to 50,000 copies/mL
  • Supplied in purified serum protein matrix identical to the composition of true clinical specimen
  • Validated using an in-house real time PCR assay

Controls should be run using the same protocols as those used to run clinical specimens.

Used to validate extraction and amplification of M. pneumoniae in human serum or plasma

Specifications

Concentration 50,000 copies/mL
Type NATtrol™ M. pneumoniae External Run Control
For Use With (Application) Used to validate extraction and amplification of M. pneumoniae in Human Serum or Plasma
Quantity 6 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.