Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Influenza A H1N1 External Run Controls
SDP

Catalog No. 22156734 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Concentration:
25 to 28 Ct
28 to 31 Ct
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
22-156-734 28 to 31 Ct
22-156-735 25 to 28 Ct
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-156-734 Supplier ZeptoMetrix Supplier No. NATFLUAH1N1ERCL
Add to Cart
Add to Cart

Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable

  • Each control pack contains 6 x 1.0mL vials of Influenza A H1N1 NATtrol™ at concentrations 25 to 31 Ct Range
  • Supplied in purified protein matrix identical to the composition of true clinical specimens
  • Designed to evaluate the performance of nucleic acid tests for determination of the presence of Influenza A H1N1 RNA

Contain intact organisms and should be run in a manner identical to that used for clinical specimens.

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Content And Storage 2°C to 8°C
Concentration 28 to 31 Ct
Detectable Analytes Influenza A H1 RNA
Type Influenza A H1N1 External Run Control
Quantity 6 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.