Learn More
ZeptoMetrix NATtrol™ Influenza A Virus
SDP
Catalog No. 22156891 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Type:
Influenza A H1 Virus
Influenza A H3 Virus
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-156-891 Influenza A H1 Virus
22-156-736 Influenza A H3 Virus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-156-891 Supplier ZeptoMetrix Supplier No. NATFLUAH1ST
Add to Cart
Add to Cart
Qualitative molecular control
  • Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
Specifications
Content And Storage 2°C to 8°C
Type Influenza A H1 Virus
Quantity 1 mL