Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix OXItek Total Glutathione Peroxidase Assay Kit
SDP

Catalog No. 22157392 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-392 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-392 Supplier ZeptoMetrix Supplier No. 0805002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

120mL of GPx-assay buffer, 45mg of GPx-reagent 1, 5mg of GPx-reagent 2, 10mL of Gpx-reagent 3, 1mL of Gpx-Reagent 4, 0.5mL of QC material.

Provides a method of quantifying the activity of total glutathione peroxidase

Glutathione peroxidase (GPx, EC No. 1.11.1.9) is an enzyme found in cytoplasmic and mitochondrial fractions of cells. GPx acts on lipid hydroperoxide (LHP) substrates that are released from membrane phospholipids by phospholipase A2. It can utilize cholesterol hydroperoxide and hydrolyzes H2O2 at low concentrations. The antioxidant enzyme catalyzes the reduction of hydrogen peroxide and hydroperoxides formed from fatty acids, thereby effectively removing toxic peroxides from living cells. It
plays the important role of protecting cells from potential damage by free radicals, formed by peroxide decomposition. The activity of GPx is coupled to glutathione reductase (GSSG-R), which maintains reduced glutathione (GSH) levels. Using glutathione (GSH) as a reducing reagent, the GPx enzymes catalyze the reduction of H2O2 and organic peroxides (R-O-O-H) to water and the corresponding stable alcohol thus inhibiting the formation of free radicals. Enzyme activity can be decreased by
negative feedback from excess substrate or from damage by oxidative modification.

Specifications

Certifications/Compliance ISO 13485
Product Type OXItek Total Glutathione Peroxidase assay Kit
Sufficient For 100 Tests
For Use With (Application) For quantifying the activity of to tal cellular Glutathione Reductase in a large number of biological specimen
Kit Contents 0.5mL QC material, 1 mL GPx-Reagent 4, 10 mL Gpx-Reagent 3, 120 mL GPx-Assay buffer, 45 mg GPx-Reagent 1, 5mg GPx reagent 2
Quantity 100 Tests
Storage Requirements 24°C to 25°C

Research purposes only. Not for in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.