Learn More
ZeptoMetrix Influenza B Virus (Massachusetts/2/12) Purified Viral Lysate
SDP
Catalog No. 22157813 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
22-157-813 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 22-157-813 Supplier ZeptoMetrix Supplier No. 0810239
Add to Cart
Add to Cart
This item is not returnable. View return policy
Can be utilized as an antigen, as a source for the purification of viral proteins, or for the detection of viral antibodies
  • Propagated in the MDCK cell line
  • Purified using sucrose density gradient ultracentrifugation, disrupted in the presence of 0.5% Triton X-100 non-ionic detergent/0.6M KCl, and heat inactivated
Specifications
Biosafety Level 2
Content And Storage Store at or below -65°C. Avoid freeze/thaw cycles.
For Use With (Application) Immunodetection of antibodies to Influenza Virus using solid-phase enzyme immunoassays (EIA); Western blot; Dot blot; Other protein-based assay
Purification Method Sucrose density gradient ultracentrifugation, disrupted in the presence of 0.5% Triton X-100 non-ionic detergent/0.6 M KCl, and heat inactivated
Quantity 1 mg
Regulatory Status RUO
For research use only. Not for use in diagnostic procedures