Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix ZeptoBind Immunoassay Stabilization Reagent
SDP

Catalog No. 22157490 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
60 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-490 1000 mL
22-157-362 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-157-490 Supplier ZeptoMetrix Supplier No. 0801184L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ensures binding of protein matrices to the plates

  • Aids in the adherence of antigens or antibodies to microwell plates
  • Liquid, ready–to–use format
  • Contains no preservatives

Use aseptic technique when removing contents.

Specifications

Content And Storage 4°C
Type Immunoassay Stabilization Reagent
Intended Use To aid in the adherence of Antigens or Antibodies to microwell plates
Quantity 1000 mL

Research purposes only. Not for in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.