Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix ZeptoBlock Immunoassay Reagent
SDP

Catalog No. 22157491 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
120 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-491 1000 mL
22-157-363 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-157-491 Supplier ZeptoMetrix Supplier No. 0801185L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Minimizes interfering substances and cross reactivity

  • Blocks nonspecific binding sites of microwell plates to which antigen or antibody has been applied
  • Liquid, ready–to–use format
  • Contains no preservatives

Use aseptic technique when removing contents.

Specifications

Content And Storage 4°C
Type Immunoassay Reagent
Intended Use To block nonspecific binding sites of microwell plates to which Antigen or Antibody has been applied
Quantity 1000 mL

Research purposes only. Not for in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.