Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix ZeptoCoat Immunoassay Stabilization Reagent
SDP

Catalog No. 22157489 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-489 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-489 Supplier ZeptoMetrix Supplier No. 0801179L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to protect and extend the shelf life of immobilized antigen and antibody matrices applied to microwell plates

  • Allows for dry storage of plates
  • Liquid, ready–to–use format
  • Contains no preservatives

Use aseptic technique when removing contents.

Specifications

Content And Storage 4°C
Type Polymeric Immunoassay Stabilization Reagent
Intended Use To protect and extend the shelf life of immobilized Antigen and Antibody matrices applied to microwell plates, increases product shelf life through protection of the Protein matrices
Quantity 1000 mL

Research purposes only. Not for in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.