Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix ZeptoGel
SDP

Catalog No. 22157365 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-365 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
May require the use of human blood or blood products.
Catalog No. 22-157-365 Supplier ZeptoMetrix Supplier No. 0801195
Add to Cart
Add to Cart
May require the use of human blood or blood products.

Separates leukocytes by taking advantage of the differential sedimentation rates of blood components.

Gentle mixing of ZeptoGel (1part) and blood (4 parts), followed by settling of the blood components for 15 to 60 minutes at 20° to 37°C, results in the formation of layers. The leukocyte fraction may be harvested by aspirating the uppermost layer.

  • Sterile
  • Isotonic
  • Pyrogen-free solution of salts plus gelatin

If ZeptoGel is gelled upon receipt, heat at 37°C for 15 minutes and then store at 15° to 30°C.

Specifications

Content And Storage 15°C to 30°C
Format Gel
For Use With (Equipment) Centrifuge
Type ZeptoGel Support Reagent
Intended Use Used to separate the Leukocytes by taking advantage of the differential sedimentation rates of Blood components
Quantity 500 mL
Sterility Sterile

Research purposes only. Not for in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.