Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Corynebacterium diphtheriae Z116, Titered
SDP

Catalog No. 21020104 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-020-104 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-020-104 Supplier ZeptoMetrix Supplier No. 0801882
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live, titered microorganisms can be used to determine a limit of detection (LOD) in diagnostic assay development or cross-reactivity studies

  • Identification confirmed by 16S sequencing
  • Purity was monitored by Gram staining and by additional culturing
  • Freezing medium contains 15% glycerol as a cryoprotectant

Specifications

Content And Storage Store at or below -65°C
For Use With (Application) Determine a limit of detection (LOD), in diagnostic assay development or crossreactivity studies
Quantity 1 mL
Form Frozen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.