Learn More
ZeptoMetrix NATtrol EBV External Run Control Strain: B95-8
SDP
Catalog No. 21020110 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Type:
External Run Control (low)
External Run Control (medium)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
21-020-110 External Run Control (low)
21-020-111 External Run Control (medium)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-020-110 Supplier ZeptoMetrix Supplier No. NATEBVERCL
Add to Cart
Add to Cart
Designed to evaluate the performance of nucleic acid tests for determination of the presence of EBV DNA
  • Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
Specifications
Type External Run Control (low)
For Use With (Application) Designed to evaluate the performance of nucleic acid tests for determination of the presence of EBV DNA, can also be used for validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specificity Evaluate the performance of Nucleic Acid