Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol EBV Linearity Panel, Strain: B95-8
SDP

Catalog No. 21020112 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-020-112 6 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-020-112 Supplier ZeptoMetrix Supplier No. NATEBVLIN
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to evaluate the performance of nucleic acid tests for determination of the presence of EBV DNA

  • Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen

Specifications

Content And Storage Store at 2° to 8°C
Formulation Purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
For Use With (Application) Evaluation the performance of nucleic acid tests for determination of the presence of EBV DNA; Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Includes 6 x 0.50 mL vials of EBV NATtrol™ at concentrations 0; 500; 5000; 50,000; 500,00; and 5,000,000 copies/mL
Quantity 6 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.