Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix RETROtek HIV-1 p24 Extended Range Kit
SDP

Catalog No. 22156774 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
5 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-774 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Not intended for use in the diagnosis or prognosis of disease or for screening and may not be used as a confirmatory test in diagnostic situations
Catalog No. 22-156-774 Supplier ZeptoMetrix Supplier No. 0801137/DEL
Not intended for use in the diagnosis or prognosis of disease or for screening and may not be used as a confirmatory test in diagnostic situations

Includes

Concentrated HIV-1 p24 Antigen Standard (0.3mL: Contains detergent disrupted, heat inactivated viral antigen at a concentration of 80ng/mL, added protein, detergent and 2-chloroacetamide, Substrate (5 tablets): Contains OPD (orthophenylenediamine-HCl)

Requires

RETRO-TEK HIV-1 p24 ELISA, Test tubes and racks for preparing specimen and standard dilutions and Adjustable micropipettes, single and multichannel

Accessory to the RETRO-TEK HIV-1 p24 Antigen ELISA

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Form Liquid
Product Type HIV Type 1 p24 Extended Range kit
For Use With (Application) Research purposes
Quantity 5 x 96 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.