Learn More
FUJIFILM Biosciences Zeranol Standard
Catalog No. 5000450346
Encompass_Preferred
Zeranol is a nonsteroidal estrogenic compound derived from zearalenone and widely studied for its occurrence in food and feed This standard is for Pesticide Residue Analysis with HPLC It is manufactured to meet the strict demands of HPLC analysis ensuring consistent and accurate results in detection and quantification workflows The material is supplied in a light-protected glass vial and should be stored refrigerated to maintain stability
Catalog No. 50-004-50346 Supplier FUJIFILM Biosciences Supplier No. 26201461
Zeranol is a nonsteroidal estrogenic compound derived from zearalenone and widely studied for its occurrence in food and feed This standard is for Pesticide Residue Analysis with HPLC It is manufactured to meet the strict demands of HPLC analysis ensuring consistent and accurate results in detection and quantification workflows The material is supplied in a light-protected glass vial and should be stored refrigerated to maintain stability