Learn More
Peak Scientific Instruments Ltd Zero Air generator with integrated air compressor
SDP
Catalog No. 5000804185
Encompass_Preferred
Zero Air generator with integrated air compressor
Up to 3.5L/min
Adjustable pressure up to 100psi
Highly efficient CAT chamber
Low power consumption, reducing power draw and operational carbon footprint
Compact design, saving valuable lab bench space
In-field upgrade to N2|ZA possible
High-quality touchscreen for operation, reminders and alerts
Auxiliary air outlet enabling support of Intura N2 or autosamplers, pyrolizers and other laboratory applications requiring clean, dry air
Advanced oil-free compressor
12-month comprehensive warranty, with the option to extend up to 5 years
Catalog No. 50-008-04185 Supplier Peak Scientific Instruments Ltd Supplier No. 3305331202
Zero Air generator with integrated air compressor
Up to 3.5L/min
Adjustable pressure up to 100psi
Highly efficient CAT chamber
Low power consumption, reducing power draw and operational carbon footprint
Compact design, saving valuable lab bench space
In-field upgrade to N2|ZA possible
High-quality touchscreen for operation, reminders and alerts
Auxiliary air outlet enabling support of Intura N2 or autosamplers, pyrolizers and other laboratory applications requiring clean, dry air
Advanced oil-free compressor
12-month comprehensive warranty, with the option to extend up to 5 years