Learn More
Climet Instruments Climet Instruments Zero Count Filter, Confirms Internal Cleanliness, ISO Class 3 Certified, 01206501R
SDP
Supplier:  Climet Instruments 01206501R
Encompass
The CLIMET INSTRUMENTS Zero Count Filter is designed for particle counter calibration and troubleshooting, ensuring accuracy in environmental monitoring. It isolates whether counting issues are internal to the instrument or from the environment. Certified to ISO Class 3 standards, this non-autoclavable filter supports flow rates of 75 LPM and 100 LPM, making it essential for reliable assessments of particle cleanliness.
Catalog No. NC1279478