Learn More
Chemglass Life Sciences Zero Dead Space Drain Valve, 2 in Beaded Pipe, GL-40 Thread, Torque Knob, For Chemglass Reactors
SDP
Supplier:  Chemglass Life Sciences CG1968GL241
Encompass_Preferred
This zero-dead-space flush-bottom glass drain valve provides complete vessel drainage and prevents solids from collecting in the vessel bottom. It is designed for pilot-plant reactors, includes a torque knob and stabilizing pin for secure operation with larger agitators, and fits a 2 in beaded pipe valve seat with a 1 in beaded pipe sidearm.
  • Flush-bottom design for complete vessel drainage.
  • 2 in beaded pipe valve seat and 1 in beaded pipe sidearm.
  • GL-40 heavy-duty glass threads.
  • PTFE plunger with perfluoro and TFE-encapsulated o-rings.
  • Perfluoro o-rings installed on the shaft for chemical resistance.
  • Anodized aluminum torque knob with laser-etched directional arrows.
  • Stabilizing pin for use with 100 L agitators to prevent shaft whipping.
  • Auto-stop mechanism when opening the valve for controlled operation.
Catalog No. 50-273-3550