Learn More
Braintree Scientific Zero Dead Volume Sterile Syringes, no connector (10/pk)
SDP
Catalog No. 501956167
Encompass
Zero Dead Volume sterile syringes offer researcher the ability to completely remove all fluid from the syringe for zero test article of blood loss. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6167 Supplier Braintree Scientific Supplier No. Z1CC
Zero Dead Volume sterile syringes offer researcher the ability to completely remove all fluid from the syringe for zero test article of blood loss. **Sold for research purposes only and not for human use**