Learn More
Braintree Scientific Zero Dead Volume Sterile Syringes with 22g connector (25/pk)
SDP
Catalog No. 501956166
Encompass
Zero Dead Volume sterile syringes offer researcher the ability to completely remove all fluid from the syringe for zero test article of blood loss. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6166 Supplier Braintree Scientific Supplier No. Z122PK
Zero Dead Volume sterile syringes offer researcher the ability to completely remove all fluid from the syringe for zero test article of blood loss. **Sold for research purposes only and not for human use**