Learn More
Braintree Scientific Zero Dead Volume Sterile Syringes,1cc minimal zero dead vol
SDP
Catalog No. 501955986
Encompass
1cc minimal zero dead volume, no connector (10/pk)Zero Dead Volume sterile syringes offer researcher the ability to completely remove all fluid from the syringe for zero test article of blood loss. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5986 Supplier Braintree Scientific Supplier No. SY110S
1cc minimal zero dead volume, no connector (10/pk)Zero Dead Volume sterile syringes offer researcher the ability to completely remove all fluid from the syringe for zero test article of blood loss. **Sold for research purposes only and not for human use**