Learn More
Mti Corporation ZERO DEFRAC PLTE 30D X1.0MM
SDP
Catalog No. NC1968957
Encompass
Zero Diffraction Plate with Cavity for XRD: 30 Dia x1.0 t mm w/ Cavity 20mm IDx0.5 mm, Si Crystal-Crystal: Si, P-type B-dopedResistivity:10-15 ohm.cmSize: 30 mm diameter x 1.0 mm Thickness,with a cavity in the center of the plate with 20mm dia. and 0.5 mm depth Zero diffraction plate is made of Silicon cut at special orientationPerfect for sample holder of powder XRD, which has no background noise from 20 to 120 degree (2θ angle Cu Ka X-Ray)
Catalog No. NC1968957 Supplier Mti Corporation Supplier No. ZEROSI30D10C1CAVITY20D
Zero Diffraction Plate with Cavity for XRD: 30 Dia x1.0 t mm w/ Cavity 20mm IDx0.5 mm, Si Crystal-Crystal: Si, P-type B-dopedResistivity:10-15 ohm.cmSize: 30 mm diameter x 1.0 mm Thickness,with a cavity in the center of the plate with 20mm dia. and 0.5 mm depth Zero diffraction plate is made of Silicon cut at special orientationPerfect for sample holder of powder XRD, which has no background noise from 20 to 120 degree (2θ angle Cu Ka X-Ray)