Learn More
Mti Corporation ZERO DIFFRACTION PLATE
SDP
Catalog No. NC1757915
Encompass
Zero Diffraction Plate for XRD: 24.6mm Dia x1.0 mm t, Si Crystal - SiZero24D10C1
Catalog No. NC1757915 Supplier Mti Corporation Supplier No. ZEROSI24D10C1US
Zero Diffraction Plate for XRD: 24.6mm Dia x1.0 mm t, Si Crystal - SiZero24D10C1