Learn More
ATLAS SCIENTIFIC Zero Dissolved Oxygen Calibration Set, 3x Zero Dissolved Oxygen Calibration Solution Pouches, 20 mL Each Pouch
SDP
Supplier:  ATLAS SCIENTIFIC ASDOP3 QTY 13
Encompass
0 mg/mL Oxygen content This product is used to confirm that Dissolved Oxygen Probe is working properly; it is used to calibrate the low end of the oxygen scale.
Catalog No. NC2825042