Learn More
ATLAS SCIENTIFIC Zero Dissolved Oxygen Calibration Set, 3x Zero Dissolved Oxygen Calibration Solution Pouches, 20ml Each Pouch
SDP
Catalog No. NC2825048
Encompass
0 mg/mL Oxygen contentThis product is used to confirm that Dissolved Oxygen Probe is working properly; it is used to calibrate the low end of the oxygen scale
Catalog No. NC2825048 Supplier ATLAS SCIENTIFIC Supplier No. ASDOP3 QTY 410
0 mg/mL Oxygen contentThis product is used to confirm that Dissolved Oxygen Probe is working properly; it is used to calibrate the low end of the oxygen scale